Tetrachloroethylene 99+%, Ultrapure for spectrophotometry

Supplier: Thermo Scientific Chemicals
Warning

32437-K2 32437-K7 32437-M1
CAAA32437-K2EA 183.28 CAD
CAAA32437-K2 CAAA32437-K7 CAAA32437-M1
Tetrachloroethylene 99+%, Ultrapure for spectrophotometry
Tetrachloroethylene

MDL: MFCD00000834 Beilstein Registry No.: 1361721


Purity: 99+% CAS No.: 127-18-4 Molecular Formula: Cl2C=CCl2 Formula Weight: 165.83 Physical Form: Liquid Density: 1.623 Refractive Index: 1.5055 Odor: Ether-like Melting Point: -22° Boiling Point: 120-121° Beilstein Reference: 1361721 Merck Reference: 14,9190 UN No.: 1897 MDL No.: MFCD00000834

Formula: C₂Cl₄
Boiling Pt: 120…121 °C
Melting Pt: –22 °C
Density: 1.623
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III
Merck Index: 14,9190
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR