Isobutanol 99+% for spectrophotometry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol

32433-K2 32433-K7 32433-M1
CAAA32433-K2EA 119.13 CAD
CAAA32433-K2 CAAA32433-K7 CAAA32433-M1
Isobutanol 99+% for spectrophotometry
Isobutanol

MDL: MFCD00004740 Beilstein Registry No.: 1730878 Notes: Meets ACS spectrophotometric requirements. Filtered through 0.2µ filters.


Purity: 99+% CAS No.: 78-83-1 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 28°(82°F) Physical Form: Liquid Density: 0.802 Refractive Index: 1.3960 Appearance: Colorless Odor: Sweet pH: 7 Melting Point: -108° Boiling Point: 107-108° Beilstein Reference: 1730878 Merck Reference: 14,5131 UN No.: 1212 MDL No.: MFCD00004740

Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 107.9
Melting Pt: -108
Density: 0.802
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3 III
Merck Index: 14,5131
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Evaporation residue 5 ppm
H2O 0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR