1-Hexanesulfonic acid sodium salt monohydrate 99%
Supplier: Thermo Scientific Chemicals
32190-09
32190-18
CAAA32190-09EA
145.2
CAD
CAAA32190-09
CAAA32190-18
1-Hexanesulfonic acid sodium salt monohydrate 99%
1-Hexanesulphonic acid sodium salt monohydrate
MDL: MFCD00149549
Beilstein Registry No.: 3727014
Purity: 99%
CAS No.: 207300-91-2
Molecular Formula: C6H13NaO3S.H2O
Formula Weight: 206.24 (188.22anhy)
Physical Form: Crystalline
Appearance: White
Beilstein Reference: 3727014
MDL No.: MFCD00149549
Formula:
C₆H₁₃NaO₃S·1H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149549 CAS Number: 207300-91-2 EINECS: 220-601-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...