Gallium tribromide, anhydrous
Supplier: Thermo Scientific Chemicals
|
Danger
|
32107-04
32107-09
CAAA32107-04EA
216.46
CAD
CAAA32107-04
CAAA32107-09
Gallium tribromide, anhydrous
Gallium tribromide
MDL: MFCD00016100
CAS No.: 13450-88-9
Molecular Formula: GaBr3
Formula Weight: 309.45
Physical Form: Crystalline
Density: 3.69
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Odorless
Melting Point: 122°
Boiling Point: 279°
UN No.: 3260
MDL No.: MFCD00016100
Formula:
Br₃Ga Boiling Pt: 279 °C Melting Pt: 122 °C Density: 3.69 Storage Temperature: Ambient |
MDL Number:
MFCD00016100 CAS Number: 13450-88-9 EINECS: 236-609-5 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...