Digallium trioxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
32102-06
32102-14
CAAA32102-06EA
130.67
CAD
CAAA32102-06
CAAA32102-14
Digallium trioxide ≥99.99% (metals basis)
Digallium trioxide
In spectroscopic analysis
Purity: 99.99%
CAS No.: 12024-21-4
Molecular Formula: Ga2O3
Formula Weight: 187.44
Physical Form: -50 Mesh Powder
Density: 6.44
Refractive Index: 1.92
Odor: Odorless
Melting Point: 1900°
Solubility: Insoluble in water. Soluble in alkalies. Very slightly soluble in hot acids
Merck Reference: 14,4346
MDL No.: MFCD00011020
In spectroscopic analysis
Formula:
Ga₂O₃ Melting Pt: 1900 °C Density: 6.44 Storage Temperature: Ambient |
MDL Number:
MFCD00011020 CAS Number: 12024-21-4 EINECS: 234-691-7 Merck Index: 14,4346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...