Phthalocyanine

Supplier: Thermo Scientific Chemicals
Warning

32073-03 32073-06 32073-14
CAAA32073-03EA 55.3 CAD
CAAA32073-03 CAAA32073-06 CAAA32073-14
Phthalocyanine
Phthalocyanine

MDL: MFCD00005085


CAS No.: 574-93-6 Molecular Formula: C32H18N8 Formula Weight: 514.52 Physical Form: Crystalline Odor: Odorless MDL No.: MFCD00005085

Formula: C₃₂H₁₈N₈
Melting Pt: 210 °C
Density: 1,447 g/cm³ (–113 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005085
CAS Number: 574-93-6
EINECS: 209-378-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR