Oleic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
31997-22
31997-06
31997-14
CAAAA31997-22EA
1068.08
CAD
CAAAA31997-22
CAAA31997-06
CAAA31997-14
Oleic acid 99%
Oleic acid
MDL: MFCD00064242
Beilstein Registry No.: 1726542
Purity: 99%
CAS No.: 112-80-1
Molecular Formula: C18H34O2
Formula Weight: 282.47
Physical Form: Liquid
Density: 0.887
Refractive Index: 1.4610
Sensitivity: Air Sensitive
Appearance: Colorless to pale yellow
Melting Point: 13-14°
Boiling Point: 286°/100mm
Beilstein Reference: 1726542
Merck Reference: 14,6828
MDL No.: MFCD00064242
Formula:
C₁₈H₃₄O₂ Boiling Pt: 286 °C (100 mmHg) Melting Pt: 13…14 °C Density: 0.887 Flash Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064242 CAS Number: 112-80-1 EINECS: 204-007-1 Merck Index: 14,6828 |
|
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...