4-Methyl-1,10-phenanthroline
Supplier: Thermo Scientific Chemicals
|
Warning
|
31888-03
31888-06
CAAA31888-03EA
124.98
CAD
CAAA31888-03
CAAA31888-06
4-Methyl-1,10-phenanthroline
4-Methyl-1,10-phenanthroline
MDL: MFCD00004978
CAS No.: 31301-28-7
Molecular Formula: C13H10N2
Formula Weight: 194.24
Physical Form: Crystalline
Melting Point: 144-145°
MDL No.: MFCD00004978
Formula:
C₁₃H₁₀N₂ Melting Pt: 144…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004978 CAS Number: 31301-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...