Iodomethane ≥99.5% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with copper
MDL: MFCD00001073
Beilstein Registry No.: 969135
Fieser: 1,682 2,274 3,202 4,341 5,447 6,384 8,342 9,308 19,215
Purity: 99.5%
CAS No.: 74-88-4
Molecular Formula: CH3I
Formula Weight: 141.94
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 2.28
Refractive Index: 1.5300
Sensitivity: Light Sensitive
Appearance: Colorless to pale yellow
Odor: Pungent
Melting Point: -67°
Boiling Point: 41-43°
Beilstein Reference: 969135
Merck Reference: 14,6087
UN No.: 2644
MDL No.: MFCD00001073
Formula:
CH₃I Boiling Pt: 41…43 °C Melting Pt: –67 °C Density: 2.28 Storage Temperature: Refrigerator |
MDL Number:
MFCD00001073 CAS Number: 74-88-4 EINECS: 200-819-5 UN: 2644 ADR: 6.1,I Merck Index: 14,6087 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...