2-Methyl-2-butene tech. 90% remainder mainly 2-methyl-1-butene, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

31853-22 31853-36
CAAA31853-22EA 111.86 CAD
CAAA31853-22 CAAA31853-36
2-Methyl-2-butene tech. 90% remainder mainly 2-methyl-1-butene, Technical Grade
2-Methyl-2-butene

MDL: MFCD00009276


Purity: 90% CAS No.: 513-35-9 Molecular Formula: CH3CH=C(CH3)2 Formula Weight: 70.13 Flash Point: -45°(-49°F) Physical Form: Liquid Density: 0.662 Refractive Index: 1.3870 Appearance: Colorless Odor: Mild Melting Point: -134° Boiling Point: 36-40° Merck Reference: 14,607 UN No.: 2460 MDL No.: MFCD00009276

Formula: C₅H₁₀
Boiling Pt: 36…40 °C
Melting Pt: –134 °C
Density: 0.662
Flash Pt: –45 °C (–49 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009276
CAS Number: 513-35-9
EINECS: 208-156-3
UN: 2460
ADR: 3,II
Merck Index: 14,607

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR