n-Undecane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Undecane
31807-18
31807-30
CAAA31807-18EA
93.06
CAD
CAAA31807-18
CAAA31807-30
n-Undecane 99%
n-Undecane
MDL: MFCD00008959
Purity: 99%
CAS No.: 1120-21-4
Molecular Formula: C11H24
Formula Weight: 156.30
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 0.74
Refractive Index: 1.4173
Appearance: Colorless
Odor: Mild
Melting Point: -26°
Boiling Point: 196°
UN No.: 2330
MDL No.: MFCD00008959
Formula:
H₃C(CH₂)₉CH₃ MW: 156.31 g/mol Boiling Pt: 196 Melting Pt: -26.7 Density: 0.74 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008959 CAS Number: 1120-21-4 EINECS: 214-300-6 UN: 2330 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...