Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dried basis) ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
31801-18
31801-30
31801-A1
CAAA31801-18EA
42.19
CAD
CAAA31801-18
CAAA31801-30
CAAA31801-A1
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dried basis) ACS
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
MDL: MFCD00004679
Beilstein Registry No.: 741883
Purity: 99.8-100.1%
CAS No.: 77-86-1
Molecular Formula: (HOCH2)3CNH2
Formula Weight: 121.14
Physical Form: Crystalline
Density: 1.353
Melting Point: 171-172°
Boiling Point: 219-220°/10mm
Beilstein Reference: 741883
Merck Reference: 14,9772
MDL No.: MFCD00004679
Formula:
NH₂C(CH₂OH)₃ Boiling Pt: 219…220 °C (10 mmHg) Melting Pt: 171…172 °C Density: 1.353 Storage Temperature: Ambient |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 EINECS: 201-064-4 Merck Index: 14,9772 |
|
Specification Test Results
Absorbance | Pass test |
H2O | 2% |
Insoluble matter | 0.005% max. |
Heavy Metals (as Pb) | 5 ppm |
Iron (Fe) | 5 ppm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...