2,2,4-Trimethylpentane 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

31787-AP 31787-K2 31787-K7 31787-M1
CAAA31787-APEA 76.31 CAD
CAAA31787-AP CAAA31787-K2 CAAA31787-K7 CAAA31787-M1
2,2,4-Trimethylpentane 99+% ACS
2,2,4-Trimethylpentane

Solvent, thinner, in determination of fuel octane numbers


Purity: 99+% CAS No.: 540-84-1 Molecular Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 Flash Point: -7°(19°F) Physical Form: Liquid Density: 0.692 Refractive Index: 1.3910 Odor: Nearly odorless Melting Point: -107° Boiling Point: 98-99° Solubility: Soluble in benzene, toluene, xylene, chloroform, ether, CS2, CCl4, and DMF Beilstein Reference: 1696876 Merck Reference: 14,5193 UN No.: 1262 MDL No.: MFCD00008943


Solvent, thinner, in determination of fuel octane numbers

Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 98…99 °C
Melting Pt: –107 °C
Density: 0.692
Flash Pt: –7 °C (19 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 14,5193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Evaporation residue 0.1%
Water-soluble titratable acid 0.003meq/g
Sulfur compounds (as S) 0.005%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR