Trifluoroacetic acid ≥99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TFA

31771-22 31771-36 31771-A4
CAAA31771-22EA 134.62 CAD
CAAA31771-22 CAAA31771-36 CAAA31771-A4
Trifluoroacetic acid ≥99.5%
Trifluoroacetic acid

MDL: MFCD00004169 Beilstein Registry No.: 742035 Fieser: 1,1219 6,613 11,557, 15,338 17,369 18,375 20,395 21,446


Purity: 99.5+% CAS No.: 76-05-1 Molecular Formula: C2HF3O2 Formula Weight: 114.02 Physical Form: Liquid Density: 1.480 Refractive Index: 1.2840 Sensitivity: Hygroscopic Appearance: Colorless Odor: Pungent Melting Point: -15° Boiling Point: 72-73° Beilstein Reference: 742035 Merck Reference: 14,9681 UN No.: 2699 MDL No.: MFCD00004169

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 72…73 °C
Melting Pt: –15 °C
Density: 1.480
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 14,9681
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR