Toluene 99,5 ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

31755-AP 31755-K2 31755-K7 31755-M1
CAAA31755-APEA 59.04 CAD
CAAA31755-AP CAAA31755-K2 CAAA31755-K7 CAAA31755-M1
Toluene 99,5 ACS
Toluene

MDL: MFCD00008512 Beilstein Registry No.: 635760 Very slightly soluble in water. Miscible with alcohol, chloroform, ether, glacial acetic acid, carbon disulfide


Purity: 99.5% CAS No.: 108-88-3 Molecular Formula: C6H5CH3 Formula Weight: 92.14 Flash Point: 4°(40°F) Physical Form: Liquid Density: 0.867 Refractive Index: 1.4967 Odor: Aromatic Melting Point: -93° Boiling Point: 111° Solubility: Very slightly soluble in water. Miscible with alcohol, chloroform, ether, glacial acetic acid, carbon disulfide Beilstein Reference: 635760 Merck Reference: 14,9529 UN No.: 1294 MDL No.: MFCD00008512

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867
Flash Pt: 4 °C (40 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 14,9529
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Liquid
Color (APHA) ≤10
Assay (GC) 99.5%
Residual water ≤0.03%
Impurity content: Sulfuric compounds (as S); ≤0.003%
Residue on evaporation ≤0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR