1,3-Diphenylguanidine 99+% primary standard
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N,N'-Diphenylguanidine, sym-Diphenylguanidine
Primary material for standardizing acids
Purity: 99+%
CAS No.: 102-06-7
Molecular Formula: (C6H5NH)2C=NH
Formula Weight: 211.27
Physical Form: Powder
Density: 1.130
Appearance: Pale Cream
Odor: Characteristic
Melting Point: 148-150°
Boiling Point: 170° dec.
Beilstein Reference: 1875653
Merck Reference: 14,3324
UN No.: 3077
MDL No.: MFCD00001758
Primary material for standardizing acids
Formula:
C₁₃H₁₃N₃ MW: 211.27 g/mol Boiling Pt: 170 °C (dec.) Melting Pt: 148…150 °C Density: 1.130 Storage Temperature: Ambient |
MDL Number:
MFCD00001758 CAS Number: 102-06-7 EINECS: 203-002-1 UN: 3077 ADR: 9,III Merck Index: 14,3324 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...