1,5-Diphenylcarbazide ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
31390-06
31390-14
31390-22
CAAA31390-06EA
52.14
CAD
CAAA31390-06
CAAA31390-14
CAAA31390-22
1,5-Diphenylcarbazide ACS
1,5-Diphenylcarbazide
MDL: MFCD00003013
Beilstein Registry No.: 752039
Soluble in hot alcohol acetone glacial acetic acid
CAS No.: 140-22-7
Molecular Formula: C13H14N4O
Formula Weight: 242.28
Physical Form: Crystalline
Sensitivity: Air and Light Sensitive
Appearance: White to cream
Melting Point: 170-176°
Solubility: Soluble in hot alcohol, acetone, glacial acetic acid
Beilstein Reference: 752039
Merck Reference: 14,3321
MDL No.: MFCD00003013
Formula:
C₁₃H₁₄N₄O Boiling Pt: 454 °C (1013 hPa) Melting Pt: 170…176 °C Density: 1.31 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003013 CAS Number: 140-22-7 EINECS: 205-403-7 Merck Index: 14,3321 |
|
Specification Test Results
Solubility in aqueous acetone | Pass test |
Residue after ignition | 0.0005 |
Sensitivity to chromate | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...