Ethyl acetate ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetic acid ethyl ester
31344-AP
31344-K2
31344-K7
31344-M1
31344-M4
CAAA31344-APEA
59.57
CAD
CAAA31344-AP
CAAA31344-K2
CAAA31344-K7
CAAA31344-M1
CAAA31344-M4
Ethyl acetate ≥99.5% ACS
Ethyl acetate
MDL: MFCD00009171
Beilstein Registry No.: 506104
Purity: 99.5+%
CAS No.: 141-78-6
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: -3°(26°F)
Physical Form: Liquid
Density: 0.902
Refractive Index: 1.3720
Appearance: Colorless
Odor: Sweet
Melting Point: -84°
Boiling Point: 77°
Beilstein Reference: 506104
Merck Reference: 14,3757
UN No.: 1173
MDL No.: MFCD00009171
Formula:
CH₃COOC₂H₅ MW: 88.11 g/mol Boiling Pt: 77.1 Melting Pt: -83 Density: 0.902 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3 II REACH: 01-2119475103-46 Merck Index: 14,3757 |
|
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.003% |
H2O | 0.002 |
Titratable acid | 0.0009 meq.g |
Substances darkened by sulfuric acid | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...