2-Furaldehyde ≥98% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Furfurol, Furfural
31305-30
31305-A1
CAAA31305-30EA
101.12
CAD
CAAA31305-30
CAAA31305-A1
2-Furaldehyde ≥98% ACS
2-Furaldehyde
MDL: MFCD00003229
Beilstein Registry No.: 105755
Purity: 98% min
CAS No.: 98-01-1
Molecular Formula: C5H4O2
Formula Weight: 96.09
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 1.159
Refractive Index: 1.5261
Sensitivity: Air Sensitive
Appearance: Yellow to brown
Odor: Almond-like
Melting Point: -38°
Boiling Point: 162°
Beilstein Reference: 105755
Merck Reference: 14,4304
UN No.: 1199
MDL No.: MFCD00003229
Formula:
C₅H₄O₂ MW: 96.09 g/mol Boiling Pt: 162 °C Melting Pt: –38 °C Density: 1.159 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003229 CAS Number: 98-01-1 EINECS: 202-627-7 UN: 1199 ADR: 6.1,II Merck Index: 14,4304 |
Specification Test Results
Titratable acid | 0.2meq/gmeq/g |
Evaporation residue | 0.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...