Dibutyl phthalate 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
31244-06
31244-14
31244-A1
31244-A5
CAAA31244-06EA
30.81
CAD
CAAA31244-06
CAAA31244-14
CAAA31244-A1
CAAA31244-A5
CAAA31244-B7
Dibutyl phthalate 99+%
Dibutyl phthalate
MDL: MFCD00009441
Beilstein Registry No.: 1914064
Very soluble in alcohol, ether, acetone, benzene
Purity: 99+%
CAS No.: 84-74-2
Molecular Formula: C16H22O4
Formula Weight: 278.35
Flash Point: 171°(340°F)
Physical Form: Liquid
Density: 1.045
Refractive Index: 1.4920
Odor: Odorless
Melting Point: -35°
Boiling Point: 339-340°
Solubility: Very soluble in alcohol, ether, acetone, benzene
Beilstein Reference: 1914064
Merck Reference: 14,3035
UN No.: 3082
MDL No.: MFCD00009441
Formula:
C₁₆H₂₂O₄ Boiling Pt: 339…340 °C Melting Pt: –35 °C Density: 1.045 Flash Pt: 171 °C (340 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009441 CAS Number: 84-74-2 EINECS: 201-557-4 UN: 3082 ADR: 9,III REACH: 01-2119493042-44 Merck Index: 14,3035 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...