1-Decene ≥96% remainder isomers
Supplier: Thermo Scientific Chemicals
|
Danger
|
31213-22
31213-36
31213-A3
CAAA31213-22EA
31.76
CAD
CAAA31213-22
CAAA31213-36
CAAA31213-A3
1-Decene ≥96% remainder isomers
1-Decene
MDL: MFCD00009577
Beilstein Registry No.: 1737236
Purity: 96%
CAS No.: 872-05-9
Molecular Formula: C10H20
Formula Weight: 140.27
Flash Point: 49°(120°F)
Physical Form: Liquid
Density: 0.741
Refractive Index: 1.4210
Appearance: Colorless
Melting Point: -66°
Boiling Point: 170-171°
Beilstein Reference: 1737236
UN No.: 3295
MDL No.: MFCD00009577
Formula:
C₁₀H₂₀ MW: 140.27 g/mol Boiling Pt: 170…171 °C Melting Pt: –66 °C Density: 0,741 Flash Pt: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009577 CAS Number: 872-05-9 EINECS: 212-819-2 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...