1,9-Decadiene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
31207-09
31207-18
CAAA31207-09EA
154.84
CAD
CAAA31207-09
CAAA31207-18
1,9-Decadiene 97%
1,9-Decadiene
MDL: MFCD00008670
Purity: 97%
CAS No.: 1647-16-1
Molecular Formula: CH2=CH(CH2)6CH=CH2
Formula Weight: 138.25
Flash Point: 42°(107°F)
Physical Form: Liquid
Density: 0.75
Refractive Index: 1.4325
Boiling Point: 169°
UN No.: 3295
MDL No.: MFCD00008670
Formula:
C₁₀H₁₈ Boiling Pt: 169 °C Melting Pt: –71.8 °C Density: 0,75 Flash Pt: 42 °C (107 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008670 CAS Number: 1647-16-1 EINECS: 216-711-6 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...