5-Chloro-1,10-phenanthroline
Supplier: Thermo Scientific Chemicals
31180-02
31180-04
CAAA31180-02EA
127.82
CAD
CAAA31180-02
CAAA31180-04
5-Chloro-1,10-phenanthroline
5-Chloro-1,10-phenanthroline
MDL: MFCD00004980
CAS No.: 4199-89-7
Molecular Formula: C12H7N2Cl
Formula Weight: 214.66
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 123°
MDL No.: MFCD00004980
Formula:
C₁₂H₇ClN₂ Melting Pt: 123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004980 CAS Number: 4199-89-7 EINECS: 224-098-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...