Iron(II) bromide, anhydrous ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
31154-06
31154-14
31154-22
CAAA31154-06EA
73.94
CAD
CAAA31154-06
CAAA31154-14
CAAA31154-22
Iron(II) bromide, anhydrous ≥98%
Iron (II) bromide
MDL: MFCD00016081
Purity: 98% min
CAS No.: 7789-46-0
Molecular Formula: Br2Fe
Formula Weight: 215.67
Physical Form: Granular
Density: 4.636
Appearance: Green to dark yellow
Odor: Odorless
Melting Point: 684°
Merck Reference: 14,4042
UN No.: 3260
MDL No.: MFCD00016081
Formula:
Br₂Fe Melting Pt: 684 °C Density: 4.636 Storage Temperature: Ambient |
MDL Number:
MFCD00016081 CAS Number: 7789-46-0 EINECS: 232-168-8 UN: 3260 ADR: 8,III Merck Index: 14,4042 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...