Iron(II) acetate Fe ≥ 29.5%, anhydrous
Supplier: Thermo Scientific Chemicals
|
Warning
|
31140-14
31140-22
CAAA31140-14EA
361.82
CAD
CAAA31140-14
CAAA31140-22
Iron(II) acetate Fe ≥ 29.5%, anhydrous
Iron (II) acetate
MDL: MFCD00058909
CAS No.: 3094-87-9
Molecular Formula: C4H6FeO4
Formula Weight: 173.94
Physical Form: Powder/Lump
Sensitivity: Air and Moisture Sensitive
Appearance: Off-white to pale brown
Odor: Characteristic
Melting Point: 190-200° dec.
MDL No.: MFCD00058909
Formula:
C₄H₆FeO₄ Melting Pt: 190…200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00058909 CAS Number: 3094-87-9 EINECS: 221-441-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...