Iron(II) bromide hydrate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Iron dibromide hydrate
31133-14
31133-22
CAAA31133-14EA
138.72
CAD
CAAA31133-14
CAAA31133-22
Iron(II) bromide hydrate
Iron (II) bromide hydrate
MDL: MFCD00016081
CAS No.: 7789-46-0
Molecular Formula: Br2Fe.xH2O (x≈5-6)
Formula Weight: 215.67(anhy)
Physical Form: Lump
Density: 3.9
Sensitivity: Air Sensitive and Hygroscopic
Appearance: Green
Odor: Odorless
Melting Point: 27°
UN No.: 3260
MDL No.: MFCD00016081
Formula:
Br₂Fe·H₂O MW: 233.67 Melting Pt: 27 °C Density: 3.9 Storage Temperature: Ambient |
MDL Number:
MFCD00016081 CAS Number: 13463-12-2 EINECS: 232-168-8 UN: 3260 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...