Pentacarbonyliron ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
As a catalyst in organic reactions, and as carbonyl iron for high-frequency coils
Purity: 99.5%
CAS No.: 13463-40-6
Molecular Formula: C5FeO5
Formula Weight: 195.90
Flash Point: -15°(5°F)
Physical Form: Liquid
Density: 1.457
Refractive Index: 1.5196
Sensitivity: Air and Light Sensitive
Appearance: Red
Odor: Pungent
Melting Point: -20°
Boiling Point: 103° (200° dec.)
Solubility: Soluble in nickel tetracarbonyl and organic solvents. Insoluble in water
Merck Reference: 14,5099
UN No.: 1994
MDL No.: MFCD00011001
As a catalyst in organic reactions, and as carbonyl iron for high-frequency coils
Formula:
C₅FeO₅ Boiling Pt: 103 °C/ 200 °C (boil./ dec.) Melting Pt: –20 °C Density: 1.457 Flash Pt: –15 °C (5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011001 CAS Number: 13463-40-6 EINECS: 236-670-8 UN: 1994 ADR: 6.1,I Merck Index: 14,5099 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...