Iron selenide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

31112-09 31112-18
CAAA31112-09EA 289.14 CAD
CAAA31112-09 CAAA31112-18
Iron selenide ≥99.9% (metals basis)
Iron selenide

MDL: MFCD00016085


Purity: 99.9% CAS No.: 1310-32-3 Molecular Formula: FeSe Formula Weight: 134.81 Physical Form: -40 Mesh Powder Sensitivity: Moisture Sensitive UN No.: 3283 MDL No.: MFCD00016085

Formula: FeSe
Melting Pt: 965 °C
Density: 4.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016085
CAS Number: 1310-32-3
EINECS: 215-177-1
UN: 3283
ADR: 6.1,III
Merck Index: 12,04103

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR