Iron(III) phosphate hydrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Iron phosphate hydrate, Iron (III) orthophosphate hydrate

31106-30 31106-A1 31106-A7
CAAA31106-30EA 82.32 CAD
CAAA31106-30 CAAA31106-A1 CAAA31106-A7
Iron(III) phosphate hydrate
Iron (III) phosphate hydrate

MDL: MFCD00016096


CAS No.: 51833-68-2 Molecular Formula: FePO4.xH2O Formula Weight: 150.82(anhy) Physical Form: Powder Appearance: Off-white

Formula: FeO₄P·xH₂O
MW: 168.83
Storage Temperature: Ambient
MDL Number: MFCD00016096
CAS Number: 51833-68-2
EINECS: 233-149-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR