Methyl ethyl ketone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
MDL: MFCD00011648
Beilstein Registry No.: 741880
Fieser: 1,678
Purity: 97%
CAS No.: 78-93-3
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -3°(26°F)
Physical Form: Liquid
Density: 0.805
Refractive Index: 1.3790
Appearance: Colorless
Odor: Characteristic
Melting Point: -87°
Boiling Point: 80°
Beilstein Reference: 741880
Merck Reference: 14,6072
UN No.: 1193
MDL No.: MFCD00011648
Formula:
C₂H₅COCH₃ MW: 72.11 g/mol Boiling Pt: 79.6 Melting Pt: -86 Density: 0.805 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3 II REACH: 01-2119457290-43 Merck Index: 14,6072 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...