1-Butanol ≥99.4% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
31068-AP
31068-K2
31068-K7
31068-M1
31068-M6
CAAA31068-APEA
64.78
CAD
CAAA31068-AP
CAAA31068-K2
CAAA31068-K7
CAAA31068-M1
CAAA31068-M6
1-Butanol ≥99.4% ACS
1-Butanol
MDL: MFCD00002964
Beilstein Registry No.: 969148
Purity: 99.4+%
CAS No.: 71-36-3
Molecular Formula: CH3(CH2)3OH
Formula Weight: 74.12
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 0.810
Refractive Index: 1.3990
Odor: Alcohol-like
pH: 7
Melting Point: -90 to -89°
Boiling Point: 117-118°
Beilstein Reference: 969148
Merck Reference: 14,1540
UN No.: 1120
MDL No.: MFCD00002964
Formula:
H₃C(CH₂)₂CH₂OH Boiling Pt: 117…118 °C Melting Pt: –90…–89 °C Density: 0.810 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III REACH: 01-2119484630-38 Merck Index: 14,1540 |
|
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.005% |
Titratable acid | 0.0008meq/g |
Carbonyl compounds (as butyraldehyde) | 0.01% |
Butyl ether | 0.2% |
H₂O | 0.1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...