1,10-Phenanthroline monohydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,10-Phenanthroline hydrate

30910-09 30910-18
CAAA30910-09EA 140.24 CAD
CAAA30910-09 CAAA30910-18
1,10-Phenanthroline monohydrate ACS
1,10-Phenanthroline monohydrate

MDL: MFCD00011678 Beilstein Registry No.: 4008096 Soluble in alcohol acetone. Slightly soluble in water


CAS No.: 5144-89-8 Molecular Formula: C12H8N2.H2O Formula Weight: 198.23 (180.21anhy) Physical Form: Crystalline Odor: Odorless Melting Point: 97-101° Beilstein Reference: 4008096 Merck Reference: 14,7214 UN No.: 2811 MDL No.: MFCD00011678

Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 97…101 °C
Density: 1.259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011678
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,7214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Suitability as redox indicator Pass Test
Suitability for determining iron Pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR