1-Pentanol 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Pentyl alcohol, 1-Hydroxy pentane, n-Amyl alcohol, Amyl alcohol, Butyl carbinol
MDL: MFCD00002977
Beilstein Registry No.: 1730975
Purity: 99+%
CAS No.: 71-41-0
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 49°(120°F)
Physical Form: Liquid
Density: 0.812
Refractive Index: 1.4103
Appearance: Colorless
Odor: Mild
Melting Point: -79°
Boiling Point: 136-138°
Beilstein Reference: 1730975
Merck Reference: 14,7118
UN No.: 1105
MDL No.: MFCD00002977
Formula:
H₃C(CH₂)₃CH₂OH MW: 88.15 g/mol Boiling Pt: 136…138 °C Melting Pt: –79 °C Density: 0.812 Flash Pt: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002977 CAS Number: 71-41-0 EINECS: 200-752-1 UN: 1105 ADR: 3 III Merck Index: 14,7118 |
|
Specification Test Results
H2O | 0.5% |
Color (APHA) | 30 |
Evaporation residue | 0.003% |
Acids and esters | 0.075meq/g |
Carbonyl (as HCHO) | 0.1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...