Tetramethylammonium hydroxide 25% (w/w) in methanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
30833-09
30833-22
30833-36
CAAA30833-09EA
57.67
CAD
CAAA30833-09
CAAA30833-22
CAAA30833-36
Tetramethylammonium hydroxide 25% (w/w) in methanol
Tetramethylammonium hydroxide
MDL: MFCD00008280
CAS No.: 75-59-2
Molecular Formula: C4H13NO
Formula Weight: 91.15
Flash Point: 12°(54°F)
Physical Form: Liquid
Density: 0.865
Appearance: Colorless to pale yellow
Odor: Like alcohol
Merck Reference: 14,9224
UN No.: 2924
MDL No.: MFCD00008280
Formula:
C₄H₁₃NO Density: 0.865 Flash Pt: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008280 CAS Number: 75-59-2 EINECS: 200-882-9 UN: 2924 ADR: 3,II Merck Index: 14,9224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...