Tetrahexylammonium perchlorate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetra-n-hexylammonium perchlorate
30830-06
30830-14
30830-22
CAAA30830-06EA
94.8
CAD
CAAA30830-06
CAAA30830-14
CAAA30830-22
Tetrahexylammonium perchlorate
Tetrahexylammonium perchlorate
MDL: MFCD00043185
CAS No.: 4656-81-9
Molecular Formula: [CH3(CH2)5]4NClO4
Formula Weight: 454.13
Physical Form: Crystalline
UN No.: 1479
MDL No.: MFCD00043185
Formula:
C₂₄H₅₂ClNO₄ MW: 454.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043185 CAS Number: 4656-81-9 EINECS: 225-095-8 UN: 1479 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...