n-Tetradecane 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetradecane
30809-14
30809-22
30809-36
CAAA30809-14EA
47.08
CAD
CAAA30809-14
CAAA30809-22
CAAA30809-36
n-Tetradecane 99+%
n-Tetradecane
MDL: MFCD00008986
Purity: 99+%
CAS No.: 629-59-4
Molecular Formula: C14H30
Formula Weight: 198.39
Flash Point: 99°(211°F)
Physical Form: Liquid
Density: 0.763
Refractive Index: 1.4290
Appearance: Colorless
Odor: Mild
Melting Point: 6°
Boiling Point: 252-254°
MDL No.: MFCD00008986
Formula:
H₃C(CH₂)₁₂CH₃ MW: 198.39 g/mol Boiling Pt: 252…254 °C Melting Pt: 6 °C Density: 0.763 Flash Pt: 99 °C (211 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008986 CAS Number: 629-59-4 EINECS: 211-096-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...