Salicylic acid 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00002439
Beilstein Registry No.: 774890
Soluble in alcohol, acetone, ether. Slightly soluble in cold water. More soluble in boiling water. Water solubility increased by Na3PO3, alkali acetates or alkali citrates
Purity: 99+%
CAS No.: 69-72-7
Molecular Formula: C7H6O3
Formula Weight: 138.12
Flash Point: 157°(315°F)
Physical Form: Powder
Density: 1.443
Melting Point: 158-160°
Boiling Point: 211°/20mm
Solubility: Soluble in alcohol, acetone, ether. Slightly soluble in cold water. More soluble in boiling water. Water solubility increased by Na3PO4, alkali acetates or alkali citrates
Beilstein Reference: 774890
Merck Reference: 14,8332
MDL No.: MFCD00002439
Formula:
(HO)C₆H₄CO₂H Boiling Pt: 211 °C (20 mmHg) Melting Pt: 158…160 °C Density: 1.443 Flash Pt: 157 °C (315 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002439 CAS Number: 69-72-7 EINECS: 200-712-3 Merck Index: 14,8332 |
|
Specification Test Results
Melting point | 158.0-161.0° |
Residue after ignition | 0.01% |
Cl | 0.001% |
SO3 | 0.003% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 2 ppm |
Substances darkened by sulfuric acid | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...