Propylene oxide 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
30765-AE
30765-K2
30765-L7
CAAA30765-AEEA
65.32
CAD
CAAA30765-AE
CAAA30765-K2
CAAA30765-L7
Propylene oxide 99+%
Propylene oxide
MDL: MFCD00005126
Beilstein Registry No.: 79763
Purity: 99+%
CAS No.: 75-56-9
Molecular Formula: C3H6O
Formula Weight: 58.08
Flash Point: -37°(-35°F)
Physical Form: Liquid
Density: 0.83
Refractive Index: 1.3660
Appearance: Colorless
Odor: Ether-like
Melting Point: -112°
Boiling Point: 34-35°
Beilstein Reference: 79763
Merck Reference: 14,7856
UN No.: 1280
MDL No.: MFCD00005126
Formula:
C₃H₆O Boiling Pt: 34…35 °C Melting Pt: –112 °C Density: 0.83 Flash Pt: –37 °C (–35 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005126 CAS Number: 75-56-9 EINECS: 200-879-2 UN: 1280 ADR: 3,I Merck Index: 14,7856 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...