Ammonium benzoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
30753-22
30753-36
30753-A3
CAAA30753-22EA
36.02
CAD
CAAA30753-22
CAAA30753-36
CAAA30753-A3
Ammonium benzoate 99%
Ammonium benzoate
MDL: MFCD00013069
Purity: 99%
CAS No.: 1863-63-4
Molecular Formula: C6H5CO2NH4
Formula Weight: 139.15
Flash Point: 110°(230°F)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White to off-white
Odor: Weak
Melting Point: 192-198° dec.
Merck Reference: 14,496
MDL No.: MFCD00013069
Formula:
C₇H₉NO₂ Melting Pt: 192…198 °C (dec.) Density: 1.26 g/cm³ (20 °C) Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013069 CAS Number: 1863-63-4 EINECS: 217-468-9 Merck Index: 14,496 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...