Aluminium iodide ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
30714-14
30714-22
CAAA30714-14EA
208.56
CAD
CAAA30714-14
CAAA30714-22
Aluminium iodide ≥95%
Aluminium iodide
Catalyst in organic synthesis
Purity: 95%
CAS No.: 7784-23-8
Molecular Formula: AlI3
Formula Weight: 407.69
Physical Form: Chunks
Density: 3.98
Appearance: White to brownish-purple
Odor: Pungent
Melting Point: 191°
Boiling Point: 360°
Solubility: Soluble in alcohol, ether, carbon disulfide
Merck Reference: 14,346
UN No.: 3260
MDL No.: MFCD00003427
Catalyst in organic synthesis
Formula:
AlI₃ Boiling Pt: 360 °C Melting Pt: 191 °C Density: 3.98 Storage Temperature: Ambient |
MDL Number:
MFCD00003427 CAS Number: 7784-23-8 EINECS: 232-054-8 UN: 3260 ADR: 8,II Merck Index: 14,346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...