Acetone ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
30698-AB
30698-AP
30698-K2
30698-K7
30698-M1
CAAA30698-ABEA
39.03
CAD
CAAA30698-AB
CAAA30698-AP
CAAA30698-K2
CAAA30698-K7
CAAA30698-M1
Acetone ≥99.5% ACS
Acetone
MDL: MFCD00008765
Beilstein Registry No.: 635680
Fieser: 2,13 3,4 6,9
Purity: 99.5+%
CAS No.: 67-64-1
Molecular Formula: C3H6O
Formula Weight: 58.08
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.791
Refractive Index: 1.3590
Appearance: Colorless
Odor: Characteristic
Melting Point: -94°
Boiling Point: 56°
Beilstein Reference: 635680
Merck Reference: 14,66
UN No.: 1090
MDL No.: MFCD00008765
Formula:
CH₃COCH₃ Boiling Pt: 56 °C Melting Pt: –94 °C Density: 0.791 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 14,66 |
|
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.001% |
Solubility in water | Pass test |
Titratable acid | 0.0003meq/g |
Titratable base | 0.0006meq/g |
Aldehyde as (HCHO) | 0.002% |
Isopropyl alcohol | 0.05% |
Methanol | 0.05% |
Substances reducing permangan | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...