2,2'-Bipyridyl 98% ACS

Supplier: Thermo Scientific Chemicals
Danger

30569-09 30569-18 30569-30
CAAA30569-09EA 59.41 CAD
CAAA30569-09 CAAA30569-18 CAAA30569-30
2,2'-Bipyridyl 98% ACS
2,2'-Bipyridyl

MDL: MFCD00006212 Beilstein Registry No.: 113089


Purity: 98% CAS No.: 366-18-7 Molecular Formula: C10H8N2 Formula Weight: 156.19 Flash Point: 121°(249°F) Physical Form: Crystalline Melting Point: 70° Boiling Point: 272-273° Beilstein Reference: 113089 Merck Reference: 14,3347 UN No.: 2811 MDL No.: MFCD00006212

Formula: C₁₀H₈N₂
Boiling Pt: 272…273 °C
Melting Pt: 70 °C
Density: 1.326 g/cm³ (–150 °C)
Flash Pt: 121 °C (249 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006212
CAS Number: 366-18-7
EINECS: 206-674-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,3347
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86bbc29edb6c5d09c058b8efcb6b9eb7f4d070c&vid=9e3a03e37045643109ae0765802e1997d30604ae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR