Strontium nitrate, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
30534-30
30534-A1
30534-D9
CAAA30534-30EA
99.22
CAD
CAAA30534-30
CAAA30534-A1
CAAA30534-D9
Strontium nitrate, anhydrous 98%
Strontium nitrate
In railroad flares, signal lights, marine signals
Purity: 98%
CAS No.: 10042-76-9
Molecular Formula: N2O6Sr
Formula Weight: 211.63
Physical Form: Granular
Density: 2.990
Appearance: White
Odor: Odorless
Melting Point: 570°
Solubility: Freely soluble in water. Slightly soluble in alcohol and acetone
Merck Reference: 14,8845
UN No.: 1507
MDL No.: MFCD00011248
In railroad flares, signal lights, marine signals
Formula:
Sr(NO₃)₂ Boiling Pt: 645 °C (1013 hPa) Melting Pt: 570 °C Density: 2.990 Storage Temperature: Ambient |
MDL Number:
MFCD00011248 CAS Number: 10042-76-9 EINECS: 233-131-9 UN: 1507 ADR: 5.1,III Merck Index: 14,8845 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...