Manganese(III) acetate hydrate ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
30519-09
30519-18
CAAA30519-09EA
135.72
CAD
CAAA30519-09
CAAA30519-18
Manganese(III) acetate hydrate ≥96%
Manganese(III) acetate hydrate
It acts as a mild and selective oxidizing agent in organic synthesis and materials science. Manganese(III) Acetate Dihydrate is also a reagent used for radical cyclizations and α-keto-acetoxylation.
Purity: 96%
CAS No.: 19513-05-4
Molecular Formula: C6H9MnO6.xH2O
Formula Weight: 232.07(anhy)
Physical Form: Crystalline
Density: 1.59
Appearance: Pink
MDL No.: MFCD00150022
Formula:
C₆H₉MnO₆·xH₂O Density: 1.59 Storage Temperature: Ambient |
MDL Number:
MFCD00150022 CAS Number: 19513-05-4 EINECS: 213-602-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...