Calcium nitrate tetrahydrate ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
30482-14
30482-22
CAAA30482-14EA
147.26
CAD
CAAA30482-14
CAAA30482-22
Calcium nitrate tetrahydrate ≥99.98% (metals basis)
Calcium nitrate tetrahydrate
MDL: MFCD00149604
Fieser: 18,84
Purity: 99.98%
CAS No.: 13477-34-4
Molecular Formula: Ca(NO3)2.4H2O
Formula Weight: 236.15 (164.09anhy)
Physical Form: Crystalline
Density: 1.82
Melting Point: 42°
Boiling Point: 132° dec.
Merck Reference: 14,1682
UN No.: 1454
MDL No.: MFCD00149604
Formula:
Ca(NO₃)₂·4H₂O MW: 236.15 g/mol Boiling Pt: 132 °C (dec.) Melting Pt: 42 Density: 1.82 Storage Temperature: Ambient |
MDL Number:
MFCD00149604 CAS Number: 13477-34-4 EINECS: 233-332-1 UN: 1454 ADR: 5.1 III Merck Index: 14,1682 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...