Barium nitrate ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
30481-14
30481-22
CAAA30481-14EA
84.85
CAD
CAAA30481-14
CAAA30481-22
Barium nitrate ≥99.95% (metals basis)
Barium nitrate
MDL: MFCD00003442
Purity: 99.95%
CAS No.: 10022-31-8
Molecular Formula: Ba(NO3)2
Formula Weight: 261.35
Physical Form: Crystalline
Density: 3.24
Melting Point: 592° dec.
Merck Reference: 14,983
UN No.: 1446
MDL No.: MFCD00003442
Formula:
Ba(NO₃)₂ Melting Pt: 592 °C (dec.) Density: 3.24 Storage Temperature: Ambient |
MDL Number:
MFCD00003442 CAS Number: 10022-31-8 EINECS: 233-020-5 UN: 1446 ADR: 5.1,II Merck Index: 14,983 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...