Trimethylphosphine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl phosphine
ampouled under argon
It is used in the preparation of organic phosphorus compounds and frequently-used phosphine ligand. It is also a highly basic ligand that forms complexes with most metals. In addition, it can react with bromomethyl-benzene to get benzyl-trimethyl-phosphonium; bromide.
Purity: 99%
CAS No.: 594-09-2
Molecular Formula: (CH3)3P
Formula Weight: 76.08
Flash Point: -30°(-22°F)
Physical Form: Liquid, Ampouled under argon
Density: 0.735
Sensitivity: Air and Moisture Sensitive
Odor: Pungent
Boiling Point: 38-40°
UN No.: 2845
MDL No.: MFCD00008510
Formula:
C₃H₉P MW: 76.08 g/mol Boiling Pt: 38…40 °C Melting Pt: –77.8 °C Density: 0.735 Flash Pt: –30 °C (–22 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008510 CAS Number: 594-09-2 EINECS: 209-823-1 UN: 2845 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...