Cobalt(II) bromide, anhydrous 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobaltous bromide

23149-18 23149-30
CAAA23149-18EA 145.2 CAD
CAAA23149-18 CAAA23149-30
Cobalt(II) bromide, anhydrous 97%
Cobalt (II) bromide

MDL: MFCD00016017 Soluble in water, methanol, ethanol, acetone, methyl acetate


Purity: 97% CAS No.: 7789-43-7 Molecular Formula: CoBr2 Formula Weight: 218.75 Physical Form: Crystalline Density: 4.909 Appearance: Green Melting Point: 678°(in N2) Solubility: Soluble in water, methanol, ethanol, acetone, methyl acetate Merck Reference: 14,2435 MDL No.: MFCD00016017

Formula: Br₂Co
MW: 218.74 g/mol
Melting Pt: 678 °C (in N₂)
Density: 4.909
Storage Temperature: Ambient
MDL Number: MFCD00016017
CAS Number: 7789-43-7
EINECS: 232-166-7
Merck Index: 14,2435

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR