Cobalt(II) iodide, anhydrous ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
23119-14
23119-22
CAAA23119-14EA
273.34
CAD
CAAA23119-14
CAAA23119-22
Cobalt(II) iodide, anhydrous ≥99.5% (metals basis)
Cobalt (II) iodide
Moisture and humidity indicator, catalyst
Purity: 99.5%
CAS No.: 15238-00-3
Molecular Formula: CoI2
Formula Weight: 312.74
Physical Form: -60 Mesh Powder
Sensitivity: Light Sensitive and Hygroscopic
Boiling Point: 570° vac.
Solubility: Soluble in water
Merck Reference: 14,2443
MDL No.: MFCD00016030
Moisture and humidity indicator, catalyst
Formula:
CoI₂ Boiling Pt: 570 °C (in vacuum) Melting Pt: 515 °C vac. Density: 5.68 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016030 CAS Number: 15238-00-3 EINECS: 239-283-2 Merck Index: 14,2443 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...