Phosphorus(III) chloride ≥99.997% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
22988-14
22988-22
CAAA22988-14EA
483.48
CAD
CAAA22988-14
CAAA22988-22
Phosphorus(III) chloride ≥99.997% (metals basis)
Phosphorus (III) chloride
MDL: MFCD00011438
Purity: 99.997%
CAS No.: 7719-12-2
Molecular Formula: PCl3
Formula Weight: 137.33
Physical Form: Liquid
Density: 1.574
Refractive Index: 1.516
Sensitivity: Moisture Sensitive
Odor: Acrid
Melting Point: -111.8°
Boiling Point: 76°
Merck Reference: 14,7358
UN No.: 1809
MDL No.: MFCD00011438
Formula:
Cl₃P Boiling Pt: 76 °C Melting Pt: –111.8 °C Density: 1.574 Storage Temperature: Ambient |
MDL Number:
MFCD00011438 CAS Number: 7719-12-2 EINECS: 231-749-3 UN: 1809 ADR: 6.1,I Merck Index: 14,7358 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...